Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016205.2 | NP_057289.1 | 4 | 1124 | ||
NR_036641.1 | 5 | 1185 | ||||
XM_011532124.2 | XP_011530426.1 | 6 | 804 | |||
XM_017008455.1 | XP_016863944.1 | 4 | 753 | |||
XM_017008456.1 | XP_016863945.1 | 4 | 1086 | |||
GenBank mRNA | AB033831.1 | - | 4 | 959 | ||
AF091434.1 | - | 4 | 1124 | |||
AF244813.1 | - | 4 | 821 | |||
AF260738.1 | - | 4 | 795 | |||
AK300480.1 | - | 3 | 610 | |||
AK304638.1 | - | 6 | 467 | |||
AK313817.1 | - | 4 | 854 | |||
AM922296.1 | - | 5 | 1005 | |||
AY358493.1 | - | 4 | 917 | |||
BC094746.1 | - | 4 | 725 | |||
BC136662.1 | - | 4 | 1045 | 9020652 |