Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018918.2 | NP_061741.1 | 1 | 1577 | ||
NM_032054.1 | NP_114443.1 | 1 | 1577 | |||
GenBank mRNA | AF152325.1 | - | 1 | 1577 | ||
AF152512.1 | - | 1 | 1577 | |||
BC104926.1 | - | 1 | 1676 | 8143929 | ||
BC113460.1 | - | 1 | 1676 | 8322512 |