Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018917.3 | NP_061740.2 | 1 | 863 | ||
NM_032053.2 | NP_114442.2 | 1 | 863 | |||
GenBank mRNA | AF152324.1 | - | 1 | 687 | ||
AF152511.1 | - | 1 | 687 |