Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018935.3 | NP_061758.1 | 1 | 2571 | ||
GenBank mRNA | AF152494.1 | - | 1 | 2341 | ||
AF217742.1 | - | 1 | 2341 | |||
AK312794.1 | - | 1 | 2573 | |||
BC038797.1 | - | 1 | 2507 | 5767743 |