Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018938.3 | NP_061761.1 | 1 | 331 | ||
GenBank mRNA | AF152497.1 | - | 1 | 82 | ||
AF217754.1 | - | 1 | 82 | |||
AK313640.1 | - | 1 | 241 | |||
BC098139.1 | - | 1 | 82 | 40008131 | ||
BC098346.1 | - | 1 | 82 | 40008130 | ||
DA231000.1 | - | 1 | 332 | |||
DA409037.1 | - | 1 | 270 |