Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018936.3 | NP_061759.1 | 1 | 1132 | ||
GenBank mRNA | AB209745.1 | - | 1 | 908 | ||
AF152495.1 | - | 1 | 948 | |||
AF217756.1 | - | 1 | 948 | |||
AK027526.1 | - | 1 | 1103 | |||
AK314642.1 | - | 1 | 1086 | |||
BC098347.1 | - | 1 | 995 | 40007951 | ||
BC098575.1 | - | 1 | 1096 | 30921306 |