Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018904.2 | NP_061727.1 | 1 | 1205 | ||
NM_031865.1 | NP_114071.1 | 1 | 1205 | |||
GenBank mRNA | AB208924.1 | - | 1 | 1266 | ||
AF152309.1 | - | 1 | 1205 | |||
AF152478.1 | - | 1 | 1205 |