Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 6 | 874 | ||
NM_001099626.1 | NP_001093096.1 | 6 | 926 | |||
NM_001099627.1 | NP_001093097.1 | 6 | 765 | |||
NM_019557.5 | NP_062457.3 | 6 | 972 | |||
GenBank mRNA | AB075880.1 | - | 6 | 802 | ||
AF173891.1 | - | 6 | 739 | |||
AK056721.1 | - | 5 | 2128 | |||
AK127164.1 | - | 2 | 1408 | |||
AK290744.1 | - | 5 | 561 | |||
AK299221.1 | - | 6 | 817 | |||
AK314297.1 | - | 6 | 802 | |||
AL512766.1 | - | 6 | 974 | |||
AY007100.1 | - | 6 | 852 | |||
BC017175.2 | - | 6 | 926 | 3162883 | ||
BX647921.1 | - | 6 | 722 | |||
KU178723.1 | - | 5 | 664 | |||
KU178724.1 | - | 5 | 687 |