Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304782.1 | NP_001291711.1 | 2 | 285 | ||
NM_019604.3 | NP_062550.2 | 7 | 833 | |||
XM_011542900.2 | XP_011541202.1 | 6 | 759 | |||
GenBank mRNA | AB209830.1 | - | 2 | 194 | ||
AF001622.1 | - | 7 | 784 | |||
AK293470.1 | - | 2 | 214 | |||
AK313578.1 | - | 7 | 831 | |||
BC070266.1 | - | 7 | 833 | 30418882 | ||
DC305734.1 | - | 2 | 285 |