Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019592.6 | NP_062538.5 | 13 | 1755 | ||
XM_011518862.1 | XP_011517164.1 | 13 | 2032 | |||
GenBank mRNA | AF265230.1 | - | 13 | 1753 | ||
AK000389.1 | - | 12 | 1646 | |||
AK000697.1 | - | 12 | 1646 | |||
AK022300.1 | - | 8 | 1027 | |||
AK022532.1 | - | 13 | 1751 | |||
AK314401.1 | - | 13 | 1710 | |||
AL834272.1 | - | 3 | 307 | |||
BC016691.1 | - | 14 | 1710 | |||
BC032618.1 | - | 9 | 1078 | |||
BC063115.1 | - | 13 | 1736 | |||
BC110584.1 | - | 13 | 1748 | 40009044 | ||
BC110585.1 | - | 13 | 1748 | 40009047 | ||
BC152309.1 | - | 13 | 1748 | 40009034 |