Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPCPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019593.3 | NP_062539.1 | 13, 14 | 1450 | ||
XM_005260758.4 | XP_005260815.1 | 14, 15 | 1601 | |||
XM_005260761.1 | XP_005260818.1 | 12, 13 | 1374 | |||
XM_006723596.3 | XP_006723659.1 | 14, 15 | 2374 | |||
GenBank mRNA | AB037855.1 | - | 13, 14 | 1441 | ||
AL833069.1 | - | 8, 9 | 2646 | |||
BC027588.2 | - | 13, 14 | 1449 | 4827346 |