Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4ENIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164501.1 | NP_001157973.1 | 16 | 2300 | ||
NM_001164502.1 | NP_001157974.1 | 14 | 1802 | |||
NM_019843.3 | NP_062817.2 | 16 | 2324 | |||
XM_005261686.2 | XP_005261743.1 | 16 | 2327 | |||
XM_005261687.2 | XP_005261744.1 | 16 | 2308 | |||
XM_005261688.2 | XP_005261745.1 | 16 | 2178 | |||
XM_006724281.3 | XP_006724344.3 | 16 | 2604 | |||
XM_011530280.1 | XP_011528582.1 | 16 | 2175 | |||
XM_011530281.2 | XP_011528583.1 | 16 | 2399 | |||
XM_017028863.1 | XP_016884352.1 | 16 | 2601 | |||
XM_017028864.1 | XP_016884353.1 | 16 | 2396 | |||
GenBank mRNA | AF240775.1 | - | 16 | 2154 | ||
AK025254.1 | - | 9 | 1055 | |||
AK074768.1 | - | 2 | 99 | |||
AK130061.1 | - | 6 | 1495 | |||
AK299136.1 | - | 8 | 1079 | |||
AK314636.1 | - | 16 | 2325 | |||
BC032941.1 | - | 14 | 1743 | 5268671 | ||
BC033028.1 | - | 16 | 2274 | 4837225 | ||
CR456386.1 | - | 15 | 2133 |