Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4ENIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164501.1 | NP_001157973.1 | 7, 8 | 1153 | ||
NM_001164502.1 | NP_001157974.1 | 5, 6 | 688 | |||
NM_019843.3 | NP_062817.2 | 7, 8 | 1177 | |||
XM_005261686.2 | XP_005261743.1 | 7, 8 | 1177 | |||
XM_005261687.2 | XP_005261744.1 | 7, 8 | 1158 | |||
XM_005261688.2 | XP_005261745.1 | 7, 8 | 1028 | |||
XM_006724281.3 | XP_006724344.3 | 7, 8 | 1454 | |||
XM_011530280.1 | XP_011528582.1 | 7, 8 | 1028 | |||
XM_011530281.2 | XP_011528583.1 | 7, 8 | 1249 | |||
XM_017028863.1 | XP_016884352.1 | 7, 8 | 1454 | |||
XM_017028864.1 | XP_016884353.1 | 7, 8 | 1249 | |||
GenBank mRNA | AF240775.1 | - | 7, 8 | 1007 | ||
AK314636.1 | - | 7, 8 | 1178 | |||
BC012734.2 | - | 7, 8 | 1030 | |||
BC032941.1 | - | 5, 6 | 629 | 5268671 | ||
BC033028.1 | - | 7, 8 | 1127 | 4837225 | ||
CR456386.1 | - | 6, 7 | 986 |