Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083947.1 | NP_001077416.1 | 8 | 951 | ||
NM_001173551.1 | NP_001167022.1 | 8 | 960 | |||
NM_001173552.1 | NP_001167023.1 | 7 | 846 | |||
NM_001290094.1 | NP_001277023.1 | 8 | 970 | |||
NM_001290096.1 | NP_001277025.1 | 8 | 881 | |||
NM_019894.3 | NP_063947.1 | 8 | 966 | |||
NR_110734.1 | 8 | 960 | ||||
XM_005271613.3 | XP_005271670.1 | 8 | 959 | |||
XM_005271614.2 | XP_005271671.1 | 8 | 924 | |||
XM_005271615.3 | XP_005271672.1 | 7 | 841 | |||
XM_011542901.2 | XP_011541203.1 | 8 | 946 | |||
XM_011542902.2 | XP_011541204.1 | 7 | 847 | |||
XM_011542903.2 | XP_011541205.1 | 8 | 960 | |||
GenBank mRNA | AF179224.1 | - | 8 | 889 | ||
AF216312.1 | - | 8 | 883 | |||
AK172766.1 | - | 8 | 822 | |||
AK290361.1 | - | 8 | 894 | |||
AK296855.1 | - | 8 | 819 | |||
AK303173.1 | - | 7 | 781 | |||
AK304113.1 | - | 8 | 934 | |||
AK304215.1 | - | 8 | 904 | |||
AK308434.1 | - | 8 | 903 | |||
AY358530.1 | - | 8 | 869 | |||
BC004855.1 | - | 8 | 911 | 3835263 | ||
BC011703.2 | - | 8 | 882 | 3610695 |