Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021253.3 | NP_067076.2 | 8 | 1288 | ||
NM_172016.2 | NP_742013.1 | 7 | 1512 | |||
GenBank mRNA | AB046381.1 | - | 7 | 1143 | ||
AK292512.1 | - | 8 | 1205 | |||
AX776007.1 | - | 7 | 1078 | |||
BC007661.2 | - | 7 | 1069 | 3051389 | ||
BC034985.1 | - | 7 | 1485 | 5268723 | ||
BT007370.1 | - | 5 | 886 |
Gene Symbol: TRIM39-RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199119.1 | NP_001186048.1 | 5 | 893 | ||
GenBank mRNA | EU012025.1 | - | 5 | 622 |