Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021253.3 | NP_067076.2 | 9 | 1439 | ||
NM_172016.2 | NP_742013.1 | 8 | 1663 | |||
GenBank mRNA | AB046381.1 | - | 8 | 1294 | ||
AK292512.1 | - | 9 | 1356 | |||
AX776007.1 | - | 8 | 1229 | |||
BC007661.2 | - | 8 | 1220 | 3051389 | ||
BC034985.1 | - | 8 | 1636 | 5268723 | ||
BT007370.1 | - | 6 | 1037 |
Gene Symbol: TRIM39-RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199119.1 | NP_001186048.1 | 6 | 1044 | ||
GenBank mRNA | EU012025.1 | - | 6 | 773 |