Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033049.3 | NP_149038.3 | 10 | 1340 | ||
GenBank mRNA | AB035807.1 | - | 10 | 1325 | ||
AF286113.1 | - | 10 | 1328 | |||
AK000070.1 | - | 8 | 716 | |||
AY358831.1 | - | 10 | 1325 |