Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286094.1 | NP_001273023.1 | 2 | 615 | ||
NM_001286095.1 | NP_001273024.1 | 3 | 264 | |||
NM_020644.2 | NP_065695.1 | 3 | 707 | |||
GenBank mRNA | AB097017.1 | - | 3 | 332 | ||
AK074677.1 | - | 3 | 325 | |||
AK300712.1 | - | 3 | 187 | |||
AK310723.1 | - | 2 | 246 | |||
AK313065.1 | - | 3 | 337 | |||
AK316216.1 | - | 2 | 246 | |||
AY359069.1 | - | 3 | 350 | |||
BC015884.1 | - | 2 | 242 | 3909465 | ||
BC040124.1 | - | 3 | 389 | 6053804 | ||
DC314550.1 | - | 3 | 264 |