Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020064.3 | NP_064448.1 | 1 | 218 | ||
GenBank mRNA | AF321618.1 | - | 1 | 26 | ||
AF325688.1 | - | 1 | 41 | |||
AK054797.1 | - | 1 | 218 | |||
BC136956.1 | - | 1 | 165 | 9020948 | ||
BC136969.1 | - | 1 | 165 | 9020961 | ||
BC144433.1 | - | 2 | 78 | 9052955 |