Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020119.3 | NP_064504.2 | 4 | 1842 | ||
NM_024625.3 | NP_078901.3 | 4 | 1842 | |||
XM_005250501.2 | XP_005250558.1 | 4 | 1843 | |||
XM_011516412.2 | XP_011514714.1 | 4 | 1843 | |||
GenBank mRNA | AK292811.1 | - | 4 | 1510 | ||
BC002935.1 | - | 1 | 555 | |||
BC025308.1 | - | 4 | 1771 | 5418915 | ||
BC033105.1 | - | 4 | 1771 | 5418956 | ||
BC040956.1 | - | 4 | 1842 | 5531412 | ||
BX571742.1 | - | 4 | 1841 | |||
BX647138.1 | - | 1 | 573 | |||
BX647974.1 | - | 4 | 1478 |