Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020125.2 | NP_064510.1 | 3 | 683 | ||
XM_005245345.2 | XP_005245402.1 | 3 | 722 | |||
XM_011509775.1 | XP_011508077.1 | 3 | 722 | |||
XM_011509776.2 | XP_011508078.1 | 3 | 584 | |||
XM_011509777.2 | XP_011508079.1 | 3 | 722 | |||
XM_011509778.2 | XP_011508080.1 | 3 | 473 | |||
XM_011509779.2 | XP_011508081.1 | 2 | 278 | |||
XM_011509780.2 | XP_011508082.2 | 2 | 290 | |||
XM_017001844.1 | XP_016857333.1 | 3 | 722 | |||
XM_017001845.1 | XP_016857334.1 | 3 | 722 | |||
XM_017001846.1 | XP_016857335.1 | 3 | 570 | |||
GenBank mRNA | AF146761.1 | - | 3 | 561 | ||
AK074669.1 | - | 3 | 622 | |||
BC109194.2 | - | 3 | 499 | 40004601 |