Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
siRNA ID
HSS125664
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
1
497
NM_001025088.1
NP_001020259.1
1
497
NM_001025089.1
NP_001020260.1
1
497
NM_020180.3
NP_064565.1
1
497
XM_005258308.3
XP_005258365.1
1
509
XM_011526079.2
XP_011524381.1
1
511
XM_011526081.2
XP_011524383.1
1
348
XM_011526083.2
XP_011524385.1
1
511
XM_011526084.2
XP_011524386.1
1
511
XM_011526085.2
XP_011524387.1
1
512
XM_011526086.2
XP_011524388.1
1
510
XM_011526087.2
XP_011524389.1
1
512
XM_011526090.2
XP_011524392.1
1
509
XM_011526091.2
XP_011524393.1
1
509
XM_011526092.2
XP_011524394.1
1
509
XM_017025840.1
XP_016881329.1
1
374
XM_017025841.1
XP_016881330.1
1
508
XM_017025842.1
XP_016881331.1
1
510
XM_017025843.1
XP_016881332.1
1
510
XM_017025844.1
XP_016881333.1
1
510
XM_017025845.1
XP_016881334.1
1
510
XM_017025846.1
XP_016881335.1
1
508
XM_017025847.1
XP_016881336.1
1
508
XM_017025848.1
XP_016881337.1
1
509
XM_017025849.1
XP_016881338.1
1
509
XM_017025850.1
XP_016881339.1
1
374
XM_017025851.1
XP_016881340.1
1
374
XM_017025852.1
XP_016881341.1
1
374
XM_017025853.1
XP_016881342.1
1
510
XM_017025854.1
XP_016881343.1
1
510
XM_017025855.1
XP_016881344.1
1
510
XM_017025856.1
XP_016881345.1
1
510
XM_017025857.1
XP_016881346.1
1
511
XM_017025858.1
XP_016881347.1
1
348
XM_017025859.1
XP_016881348.1
1
348
XM_017025860.1
XP_016881349.1
1
509
XM_017025861.1
XP_016881350.1
1
374
XM_017025862.1
XP_016881351.1
1
374
XM_017025863.1
XP_016881352.1
1
356
XM_017025864.1
XP_016881353.1
1
348
XM_017025865.1
XP_016881354.1
1
374
GenBank mRNA
AB209774.1
-
1
291
AF248650.1
-
1
236
AF329265.1
-
1
101
AK074596.1
-
1
229
AK295009.1
-
1
111
BC001946.1
-
1
258
2820541
BC004167.2
-
1
258
2820541
BC045711.1
-
1
184
5261155
BQ637723.1
-
1
504
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline