Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128427.2 | NP_001121899.1 | 4 | 747 | ||
NM_001128428.2 | NP_001121900.1 | 4 | 747 | |||
NM_020116.4 | NP_064501.2 | 4 | 750 | |||
XM_011532126.1 | XP_011530428.1 | 4 | 716 | |||
GenBank mRNA | AB033089.1 | - | 4 | 677 | ||
AK315152.1 | - | 4 | 718 | |||
AL137695.1 | - | 1 | 94 | |||
BC036502.2 | - | 4 | 751 | 5265170 | ||
BC073978.1 | - | 4 | 450 |