Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020122.4 | NP_064507.3 | 4, 5 | 768 | ||
XM_006712052.3 | XP_006712115.1 | 4, 5 | 800 | |||
XM_011532990.1 | XP_011531292.1 | 4, 5 | 690 | |||
XM_017004511.1 | XP_016860000.1 | 3, 4 | 404 | |||
GenBank mRNA | AB083199.1 | - | 4, 5 | 477 | ||
AF155652.1 | - | 4, 5 | 477 | |||
AK023403.1 | - | 4, 5 | 447 | |||
AK314761.1 | - | 4, 5 | 543 | |||
AL122115.1 | - | 2, 3 | 121 | |||
BC000178.2 | - | 4, 5 | 567 | 3048124 | ||
BU844982.1 | - | 4, 5 | 752 |