Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020123.3 | NP_064508.3 | 13 | 1791 | ||
XM_011539976.2 | XP_011538278.1 | 13 | 1871 | |||
XM_011539977.1 | XP_011538279.1 | 13 | 1920 | |||
GenBank mRNA | AF116347.1 | - | 13 | 1541 | ||
AF160213.1 | - | 13 | 1584 | |||
AF269150.1 | - | 13 | 1791 | |||
AK000756.1 | - | 11 | 1194 | |||
AK027438.1 | - | 8 | 841 | |||
AK027788.1 | - | 12 | 1447 | |||
AK074824.1 | - | 10 | 1082 | |||
AK075093.1 | - | 8 | 829 | |||
AK075451.1 | - | 2 | 194 | |||
AK297721.1 | - | 9 | 744 | |||
AK314348.1 | - | 13 | 1757 | |||
AY358819.1 | - | 12 | 1461 | |||
BC020959.1 | - | 12 | 1371 | |||
BC136788.1 | - | 13 | 1791 | 9020778 | ||
BC136789.1 | - | 13 | 1791 | 9020779 |