Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020135.2 | NP_064520.2 | 3, 4 | 1458 | ||
NM_130395.2 | NP_569079.1 | 3, 4 | 1383 | |||
GenBank mRNA | AB056152.1 | - | 3, 4 | 1459 | ||
AB209723.1 | - | 3, 4 | 1211 | |||
AF218313.1 | - | 3, 4 | 845 | |||
AK026179.1 | - | 2, 3 | 363 | |||
AK094823.1 | - | 3, 4 | 1418 | |||
AK223593.1 | - | 3, 4 | 1392 | |||
AK315047.1 | - | 3, 4 | 1550 | |||
AK315471.1 | - | 3, 4 | 1352 | |||
BC018923.2 | - | 3, 4 | 1344 | 4109718 |