Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020144.4 | NP_064529.4 | 1 | 1603 | ||
GenBank mRNA | AF218840.1 | - | 1 | 1524 | ||
AK312788.1 | - | 1 | 1635 | |||
BC036653.2 | - | 1 | 1603 | 5269035 |