Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020151.3 | NP_064536.2 | 8 | 1415 | ||
XM_011511499.1 | XP_011509801.1 | 8 | 1032 | |||
GenBank mRNA | AF270647.1 | - | 8 | 1342 | ||
AK023234.1 | - | 7 | 674 | |||
AK054615.1 | - | 8 | 781 | |||
AK094925.1 | - | 2 | 987 | |||
AK291190.1 | - | 8 | 797 | |||
BC003028.1 | - | 8 | 1398 | |||
BC007894.2 | - | 9 | 1173 | 4134933 | ||
BC008894.2 | - | 8 | 1398 | 3833465 | ||
BC009998.2 | - | 9 | 1173 | 4128869 | ||
BC012774.2 | - | 8 | 1365 | 3842611 | ||
BC012793.2 | - | 8 | 1379 | 4080200 | ||
BC013279.2 | - | 8 | 1398 | 3830320 | ||
BC014076.2 | - | 8 | 1375 | 4127007 | ||
BC014274.2 | - | 8 | 1394 | 3677198 | ||
BC032106.1 | - | 8 | 1389 | 4643387 |