Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162483.1 | NP_001155955.1 | 1 | 259 | ||
NM_020161.3 | NP_064546.3 | 1 | 259 | |||
GenBank mRNA | AL359944.1 | - | 1 | 111 | ||
BC131618.1 | - | 1 | 97 | 40116809 | ||
CR749679.1 | - | 1 | 111 |