Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020177.2 | NP_064562.1 | 3 | 1283 | ||
XM_005272035.4 | XP_005272092.1 | 3 | 1285 | |||
XM_017009647.1 | XP_016865136.1 | 4 | 1369 | |||
GenBank mRNA | AB058688.1 | - | 2 | 811 | ||
AF391093.1 | - | 2 | 721 | |||
AK315803.1 | - | 3 | 1061 | |||
AL365415.1 | - | 3 | 1021 | |||
AY249188.1 | - | 2 | 721 | |||
BC028369.2 | - | 3 | 1285 | 4826273 |