Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020207.4 | NP_064592.2 | 16 | 3735 | ||
XM_011518641.2 | XP_011516943.1 | 15 | 3582 | |||
XM_011518644.2 | XP_011516946.1 | 15 | 3527 | |||
XM_011518645.2 | XP_011516947.1 | 16 | 3735 | |||
XM_011518646.2 | XP_011516948.1 | 16 | 3735 | |||
XM_011518647.1 | XP_011516949.1 | 16 | 3735 | |||
XM_011518648.2 | XP_011516950.1 | 7 | 1807 | |||
XM_011518650.2 | XP_011516952.1 | 16 | 3054 | |||
XM_017014707.1 | XP_016870196.1 | 16 | 3722 | |||
GenBank mRNA | AK308131.1 | - | 1 | 770 | ||
AL133663.1 | - | 11 | 2335 | |||
BC022957.1 | - | 1 | 164 | 4995030 | ||
BQ013632.1 | - | 3 | 543 | |||
CA425791.1 | - | 3 | 537 |