Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020209.3 | NP_064594.3 | 1 | 1706 | ||
GenBank mRNA | AK055673.1 | - | 1 | 645 | ||
AK056268.1 | - | 1 | 1707 | |||
BC007206.1 | - | 1 | 468 | 3628187 |