Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 10, 11 | 1117 | ||
NR_024265.1 | 10, 11 | 1260 | ||||
XM_006713705.3 | XP_006713768.1 | 10, 11 | 1069 | |||
XM_017006878.1 | XP_016862367.1 | 10, 11 | 1232 | |||
XM_017006879.1 | XP_016862368.1 | 11, 12 | 1378 | |||
XM_017006880.1 | XP_016862369.1 | 11, 12 | 1235 | |||
GenBank mRNA | AK027012.1 | - | 1, 2 | 46 | ||
AK075444.1 | - | 10, 11 | 1053 | |||
AK126736.1 | - | 2, 3 | 2746 | |||
AK222937.1 | - | 10, 11 | 1077 | |||
AK295984.1 | - | 10, 11 | 1235 | |||
AK315367.1 | - | 10, 11 | 1092 | |||
AY298903.1 | - | 10, 11 | 1103 | |||
AY358581.1 | - | 10, 11 | 1060 | |||
BC030614.2 | - | 10, 11 | 1103 | 4796951 | ||
BC048810.1 | - | 10, 11 | 1061 | 5243487 |