Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 5 | 647 | ||
NR_024265.1 | 5 | 790 | ||||
XM_006713705.3 | XP_006713768.1 | 5 | 599 | |||
XM_017006878.1 | XP_016862367.1 | 5 | 762 | |||
XM_017006879.1 | XP_016862368.1 | 6 | 908 | |||
XM_017006880.1 | XP_016862369.1 | 6 | 765 | |||
GenBank mRNA | AF168711.1 | - | 5 | 598 | ||
AK075444.1 | - | 5 | 583 | |||
AK222937.1 | - | 5 | 607 | |||
AK295984.1 | - | 5 | 765 | |||
AK301002.1 | - | 6 | 906 | |||
AK315367.1 | - | 5 | 622 | |||
AY298903.1 | - | 5 | 633 | |||
AY358581.1 | - | 5 | 590 | |||
BC030614.2 | - | 5 | 633 | 4796951 | ||
BC048810.1 | - | 5 | 591 | 5243487 |