Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 4, 5 | 806 | ||
NM_020234.5 | NP_064619.2 | 5, 6 | 934 | |||
XM_017022419.1 | XP_016877908.1 | 4, 5 | 853 | |||
XM_017022420.1 | XP_016877909.1 | 5, 6 | 1715 | |||
XM_017022421.1 | XP_016877910.1 | 5, 6 | 946 | |||
XM_017022422.1 | XP_016877911.1 | 4, 5 | 1587 | |||
XM_017022427.1 | XP_016877916.1 | 5, 6 | 871 | |||
XM_017022428.1 | XP_016877917.1 | 5, 6 | 876 | |||
XM_017022429.1 | XP_016877918.1 | 5, 6 | 1822 | |||
XM_017022430.1 | XP_016877919.1 | 5, 6 | 1025 | |||
XM_017022431.1 | XP_016877920.1 | 4, 5 | 467 | |||
GenBank mRNA | AF168717.1 | - | 4, 5 | 794 | ||
AK290184.1 | - | 4, 5 | 775 | |||
BC032535.2 | - | 5, 6 | 874 | 5527713 | ||
BC093073.1 | - | 5, 6 | 864 | 30326731 |