Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020244.2 | NP_064629.2 | 7 | 1272 | ||
XM_011538574.1 | XP_011536876.1 | 6 | 1107 | |||
XM_011538575.1 | XP_011536877.1 | 7 | 969 | |||
GenBank mRNA | AF047431.1 | - | 7 | 842 | ||
AF111803.1 | - | 6 | 707 | |||
AF195623.1 | - | 7 | 1219 | |||
AF195624.1 | - | 8 | 1246 | |||
AK075211.1 | - | 7 | 1239 | |||
AK093067.1 | - | 8 | 1132 | |||
AK226127.1 | - | 8 | 973 | |||
AY166717.1 | - | 3 | 347 | |||
AY166718.1 | - | 3 | 347 | |||
AY280609.1 | - | 5 | 522 | |||
AY280610.1 | - | 5 | 530 | |||
AY294627.1 | - | 5 | 530 | |||
BC020819.1 | - | 7 | 1226 | 4696613 | ||
BC050429.1 | - | 7 | 1265 | 6473945 |