Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318785.1 | NP_001305714.1 | 10 | 1172 | ||
NM_020320.4 | NP_064716.2 | 11 | 1007 | |||
NR_134857.1 | 11 | 1007 | ||||
XM_011535948.2 | XP_011534250.1 | 11 | 992 | |||
XM_011535949.2 | XP_011534251.1 | 11 | 992 | |||
XM_017011072.1 | XP_016866561.1 | 11 | 1173 | |||
XM_017011073.1 | XP_016866562.1 | 12 | 1391 | |||
XM_017011074.1 | XP_016866563.1 | 11 | 994 | |||
XM_017011075.1 | XP_016866564.1 | 10 | 1117 | |||
XM_017011076.1 | XP_016866565.1 | 12 | 1335 | |||
XM_017011077.1 | XP_016866566.1 | 10 | 894 | |||
XM_017011078.1 | XP_016866567.1 | 10 | 938 | |||
XM_017011079.1 | XP_016866568.1 | 11 | 1173 | |||
XM_017011080.1 | XP_016866569.1 | 12 | 1335 | |||
XM_017011081.1 | XP_016866570.1 | 10 | 938 | |||
GenBank mRNA | AK023550.1 | - | 11 | 950 | ||
AK093934.1 | - | 11 | 1380 | |||
AK315669.1 | - | 11 | 944 | |||
BC010420.1 | - | 11 | 978 | 3613406 | ||
BC105784.1 | - | 10 | 1111 |