Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020648.5 | NP_065699.1 | 3 | 346 | ||
GenBank mRNA | AF196834.1 | - | 3 | 303 | ||
AF294628.1 | - | 2 | 168 | |||
AJ297391.1 | - | 3 | 297 | |||
AK027629.1 | - | 3 | 346 | |||
AK075550.1 | - | 3 | 346 | |||
AK315751.1 | - | 3 | 231 | |||
BC020490.1 | - | 3 | 260 | 3889103 | ||
DA735800.1 | - | 3 | 346 |