Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020648.5 | NP_065699.1 | 5 | 722 | ||
GenBank mRNA | AF196834.1 | - | 5 | 679 | ||
AF294628.1 | - | 4 | 544 | |||
AJ297391.1 | - | 5 | 673 | |||
AK027629.1 | - | 7 | 880 | |||
AK075550.1 | - | 5 | 722 | |||
AK315751.1 | - | 5 | 607 | |||
BC020490.1 | - | 5 | 636 | 3889103 | ||
DA735800.1 | - | 5 | 723 |