Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 6 | 839 | ||
XM_011517570.2 | XP_011515872.1 | 5 | 1253 | |||
XM_017013646.1 | XP_016869135.1 | 6 | 481 | |||
XM_017013647.1 | XP_016869136.1 | 6 | 1568 | |||
GenBank mRNA | AF221594.1 | - | 6 | 796 | ||
AY044833.1 | - | 5 | 672 | |||
BC033684.1 | - | 6 | 799 | 5193403 |