Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020371.2 | NP_065104.1 | 2 | 477 | ||
XM_011521818.2 | XP_011520120.1 | 3 | 660 | |||
XM_011521819.2 | XP_011520121.1 | 2 | 703 | |||
XM_011521820.1 | XP_011520122.1 | 2 | 178 | |||
GenBank mRNA | AF283508.1 | - | 2 | 473 | ||
BC010488.1 | - | 2 | 471 | 2900586 | ||
BC063533.1 | - | 2 | 461 | 4425400 | ||
BU855198.1 | - | 551 |