Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020371.2 | NP_065104.1 | 2, 3 | 512 | ||
XM_011521818.2 | XP_011520120.1 | 3, 4 | 695 | |||
XM_011521819.2 | XP_011520121.1 | 2, 3 | 738 | |||
XM_011521820.1 | XP_011520122.1 | 2, 3 | 213 | |||
GenBank mRNA | AF283508.1 | - | 2, 3 | 508 | ||
BC010488.1 | - | 2, 3 | 506 | 2900586 | ||
BC063533.1 | - | 2, 3 | 496 | 4425400 | ||
BU855198.1 | - | 586 |