Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRASLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020386.4 | NP_065119.2 | 2 | 521 | ||
XM_011513034.1 | XP_011511336.1 | 2 | 521 | |||
XM_011513035.2 | XP_011511337.1 | 2 | 521 | |||
XM_017006923.1 | XP_016862412.1 | 3 | 402 | |||
GenBank mRNA | AB030816.1 | - | 2 | 326 | ||
AB510981.1 | - | 1 | 115 | |||
AY251533.1 | - | 1 | 128 | |||
BC048095.1 | - | 2 | 524 | 4830228 | ||
BX098792.1 | - | 3 | 552 |