Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020397.3 | NP_065130.1 | 8 | 1060 | ||
NM_153498.3 | NP_705718.1 | 8 | 1060 | |||
XM_006717481.3 | XP_006717544.1 | 8 | 1324 | |||
XM_006717482.3 | XP_006717545.1 | 8 | 1089 | |||
XM_006717483.3 | XP_006717546.1 | 8 | 1051 | |||
XM_011519591.2 | XP_011517893.1 | 8 | 1497 | |||
XM_011519592.2 | XP_011517894.1 | 7 | 1689 | |||
XM_011519593.2 | XP_011517895.1 | 7 | 1737 | |||
XM_011519594.2 | XP_011517896.1 | 6 | 678 | |||
XM_011519595.2 | XP_011517897.1 | 7 | 1551 | |||
XM_017016438.1 | XP_016871927.1 | 9 | 1355 | |||
XM_017016439.1 | XP_016871928.1 | 10 | 1186 | |||
GenBank mRNA | AB081726.1 | - | 8 | 1016 | ||
AF286366.1 | - | 8 | 871 | |||
AK289393.1 | - | 9 | 871 | |||
BC035745.1 | - | 8 | 877 | 5748490 |