Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321859.1 | NP_001308788.1 | 7, 8 | 3025 | ||
NM_001321860.1 | NP_001308789.1 | 7, 8 | 3095 | |||
NM_001321861.1 | NP_001308790.1 | 7, 8 | 3058 | |||
NM_001321862.1 | NP_001308791.1 | 7, 8 | 3045 | |||
NM_001321863.1 | NP_001308792.1 | 7, 8 | 2995 | |||
NM_001321904.1 | NP_001308833.1 | 8, 9 | 3473 | |||
NM_007008.2 | NP_008939.1 | 5, 6 | 715 | |||
NM_020532.4 | NP_065393.1 | 7, 8 | 3778 | |||
NM_153828.2 | NP_722550.1 | 5, 6 | 1321 | |||
NM_207520.1 | NP_997403.1 | 6, 7 | 1378 | |||
NM_207521.1 | NP_997404.1 | 7, 8 | 3036 | |||
NR_135829.1 | 5, 6 | 601 | ||||
NR_135830.1 | 5, 6 | 579 | ||||
XM_005264434.3 | XP_005264491.1 | 8, 9 | 3172 | |||
XM_017004518.1 | XP_016860007.1 | 8, 9 | 3211 | |||
XM_017004519.1 | XP_016860008.1 | 8, 9 | 3328 | |||
GenBank mRNA | AB015639.1 | - | 5, 6 | 1089 | ||
AB020693.1 | - | 7, 8 | 3614 | |||
AB040462.1 | - | 7, 8 | 3724 | |||
AB040463.1 | - | 5, 6 | 1267 | |||
AB073351.1 | - | 5, 6 | 1450 | |||
AF063601.1 | - | 5, 6 | 2038 | |||
AF077050.1 | - | 5, 6 | 714 | |||
AF087901.1 | - | 5, 6 | 606 | |||
AF125103.1 | - | 5, 6 | 715 | |||
AF132047.1 | - | 5, 6 | 1202 | |||
AF132048.1 | - | 5, 6 | 621 | |||
AF148537.1 | - | 7, 8 | 3621 | |||
AF148538.1 | - | 5, 6 | 1164 | |||
AF177332.1 | - | 5, 6 | 620 | |||
AF320999.1 | - | 6, 7 | 2784 | |||
AF333336.1 | - | 7, 8 | 3040 | |||
AJ251384.1 | - | 5, 6 | 1023 | |||
AJ251385.1 | - | 5, 6 | 501 | |||
AK075039.1 | - | 5, 6 | 1255 | |||
AK129806.1 | - | 5, 6 | 1180 | |||
AK130812.1 | - | 5, 6 | 1082 | |||
AK172726.1 | - | 3, 4 | 145 | |||
AY102276.1 | - | 5, 6 | 735 | |||
AY102277.1 | - | 5, 6 | 1267 | |||
AY102278.1 | - | 6, 7 | 1324 | |||
AY102279.1 | - | 7, 8 | 3724 | |||
AY123245.1 | - | 7, 8 | 3037 | |||
AY123246.1 | - | 7, 8 | 3095 | |||
AY123247.1 | - | 7, 8 | 3058 | |||
AY123248.1 | - | 7, 8 | 3044 | |||
AY123249.1 | - | 7, 8 | 3005 | |||
AY123250.1 | - | 7, 8 | 2995 | |||
BC001035.2 | - | 5, 6 | 686 | |||
BC007109.1 | - | 5, 6 | 612 | |||
BC010737.1 | - | 5, 6 | 1028 | |||
BC012619.1 | - | 5, 6 | 1178 | |||
BC014366.1 | - | 5, 6 | 612 | |||
BC016165.1 | - | 6, 7 | 1327 | |||
BC026788.1 | - | 5, 6 | 1209 | |||
BC068991.1 | - | 5, 6 | 1271 | |||
BC071848.1 | - | 6, 7 | 1009 | |||
BC150182.1 | - | 7, 8 | 3728 | 40148210 | ||
BC152425.1 | - | 7, 8 | 3614 | 8862539 | ||
BC152555.1 | - | 7, 8 | 3728 | 40148984 | ||
CN429712.1 | - | 5, 6 | 550 | |||
DQ778739.1 | - | 5, 6 | 501 |