Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020423.6 | NP_065156.5 | 13 | 2268 | ||
NM_181093.3 | NP_851607.2 | 14 | 2430 | |||
XM_006711465.1 | XP_006711528.1 | 14 | 2379 | |||
XM_011509801.1 | XP_011508103.1 | 14 | 2519 | |||
XM_011509802.1 | XP_011508104.1 | 14 | 2379 | |||
XM_011509803.1 | XP_011508105.1 | 13 | 2282 | |||
XM_017001862.1 | XP_016857351.1 | 13 | 2217 | |||
XM_017001863.1 | XP_016857352.1 | 13 | 2357 | |||
XM_017001864.1 | XP_016857353.1 | 13 | 2217 | |||
XM_017001865.1 | XP_016857354.1 | 13 | 2316 | |||
GenBank mRNA | AF540957.1 | - | 12 | 2098 | ||
AK292507.1 | - | 14 | 2376 | |||
AK304481.1 | - | 8 | 1860 | |||
AL117233.1 | - | 13 | 2151 | |||
AY144493.1 | - | 13 | 2260 | |||
BC014662.1 | - | 13 | 2205 | 4852719 |
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005245317.4 | XP_005245374.1 | 24 | 3577 | ||
XM_011509735.2 | XP_011508037.1 | 21 | 3187 | |||
XM_017001724.1 | XP_016857213.1 | 23 | 3449 | |||
XM_017001725.1 | XP_016857214.1 | 22 | 3310 | |||
GenBank mRNA | AK023532.1 | - | 14 | 2440 |