Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145056.1 | NP_001138528.1 | 22 | 2134 | ||
NM_020428.3 | NP_065161.3 | 22 | 2193 | |||
XM_005259997.1 | XP_005260054.1 | 23 | 2318 | |||
XM_005259999.1 | XP_005260056.1 | 23 | 2259 | |||
GenBank mRNA | AF070636.1 | - | 3 | 194 | ||
AK027519.1 | - | 22 | 2078 | |||
AK075512.1 | - | 11 | 1095 | |||
AK126550.1 | - | 10 | 884 | |||
AK294118.1 | - | 6 | 487 | |||
AK314582.1 | - | 22 | 2135 | |||
AL832517.1 | - | 12 | 1621 | |||
AL832978.1 | - | 19 | 1877 | |||
BC010617.2 | - | 19 | 1884 | |||
BC040556.1 | - | 22 | 2135 | 5268313 | ||
BC042692.1 | - | 22 | 2135 | |||
FN668568.1 | - | 22 | 2193 |