Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 13, 14 | 1284 | ||
GenBank mRNA | AK125159.1 | - | 13, 14 | 1216 | ||
BC136613.1 | - | 13, 14 | 1166 | 9020603 | ||
BC136614.1 | - | 13, 14 | 1163 | 9020604 |