Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135917.1 | NP_001129389.1 | 6 | 544 | ||
NM_020438.4 | NP_065171.2 | 7 | 673 | |||
NR_072983.1 | 5 | 459 | ||||
GenBank mRNA | AK290713.1 | - | 7 | 673 | ||
AK293627.1 | - | 6 | 535 | |||
AY189675.1 | - | 7 | 676 | |||
BC009493.2 | - | 7 | 643 | |||
BC033686.1 | - | 7 | 650 | 5195127 | ||
BC040152.1 | - | 5 | 429 |