Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127257.1 | NP_001120729.1 | 7 | 1934 | ||
NM_020342.2 | NP_065075.1 | 7 | 1885 | |||
XM_005246689.4 | XP_005246746.2 | 7 | 1892 | |||
XM_011511504.2 | XP_011509806.1 | 7 | 1799 | |||
XM_011511505.2 | XP_011509807.1 | 8 | 1963 | |||
XM_011511506.2 | XP_011509808.1 | 7 | 4622 | |||
XM_011511507.2 | XP_011509809.1 | 7 | 1892 | |||
XM_017004522.1 | XP_016860011.1 | 7 | 2924 | |||
GenBank mRNA | AB033091.1 | - | 7 | 1798 | ||
AK291241.1 | - | 7 | 1797 | |||
AK294771.1 | - | 8 | 981 | |||
BC073909.1 | - | 3 | 310 | |||
BC101516.1 | - | 7 | 1794 | 8069022 | ||
BC112223.1 | - | 7 | 1806 | 8327691 | ||
CR749813.1 | - | 7 | 1785 |